Assay Details
| Gene Symbol: Wdr70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081402.1 | NP_001074871.1 | 3 - 4 | 236 | 108 | |
| GenBank mRNA | AK159714.1 | - | 3 - 4 | 218 | 108 | |
| AK160933.1 | - | 3 - 4 | 171 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Wdr70
Genomic Map