Assay Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029934.1 | NP_001025105.1 | 26 - 27 | 3435 | 81 | |
| GenBank mRNA | BC040268.1 | - | 3 - 4 | 400 | 81 | |
| BC051399.1 | - | 3 - 4 | 473 | 81 | ||
| BC058994.1 | - | 7 - 8 | 976 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp32
Genomic Map