Assay Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029934.1 | NP_001025105.1 | 27 - 28 | 3623 | 94 | |
| GenBank mRNA | BC040268.1 | - | 4 - 5 | 588 | 94 | |
| BC051399.1 | - | 4 - 5 | 661 | 94 | ||
| BC058994.1 | - | 8 - 9 | 1164 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp32
Genomic Map