Assay Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029934.1 | NP_001025105.1 | 29 - 30 | 3834 | 67 | |
| GenBank mRNA | BC040268.1 | - | 6 - 7 | 799 | 67 | |
| BC051399.1 | - | 6 - 7 | 872 | 67 | ||
| BC058994.1 | - | 10 - 11 | 1375 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp32
Genomic Map