Assay Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029934.1 | NP_001025105.1 | 32 - 33 | 4736 | 131 | |
| GenBank mRNA | AK140739.1 | - | 1 - 2 | 109 | 131 | |
| BC040268.1 | - | 9 - 10 | 1701 | 131 | ||
| BC051399.1 | - | 9 - 10 | 1774 | 131 | ||
| BC058994.1 | - | 13 - 14 | 2277 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp32
Genomic Map