Assay Details
| Gene Symbol: Usp32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029934.1 | NP_001025105.1 | 13 - 14 | 1620 | 116 | |
| GenBank mRNA | AK148221.1 | - | 13 - 14 | 1620 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp32
Genomic Map