| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100031364.mm.1 |
NM_001005223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031364.mm.1 |
NM_001005223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031364.mm.1 |
TC1100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100031360.mm.1 |
NM_001005223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031360.mm.1 |
NM_001005223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031360.mm.1 |
TC1100003421.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100031363.mm.1 |
NM_001005223.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031363.mm.1 |
NM_001005223.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100031363.mm.1 |
TC1100003421.mm.1 |
MTA TC |