Assay Details
| Gene Symbol: Znhit3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005223.2 | NP_001005223.1 | 4 - 5 | 288 | 86 | |
| GenBank mRNA | AK002888.1 | - | 4 - 5 | 280 | 86 | |
| AK003721.1 | - | 4 - 5 | 287 | 86 | ||
| AK004259.1 | - | 4 - 5 | 290 | 86 | ||
| AK014350.1 | - | 4 - 5 | 299 | 86 | ||
| BC089554.1 | - | 4 - 5 | 376 | 30300471 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980352 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 170399 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 634860 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 956928 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P125978 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980358 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980344 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980350 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980342 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980384 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980347 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980385 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980351 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980375 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980368 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980387 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980366 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980372 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27980378 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P125978 | - | - |
| Whole Mouse Genome G4122A | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | NM_001005223.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK002888.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK003721.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK004259.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P654618 | AK014350.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115526 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129222 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P125978 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08982977 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09364777 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09040080 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08982976 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980353 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980351 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27980352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980315 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980295 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980317 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980302 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27980352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09187846 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980297 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980310 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09063024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09136665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09729350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980342 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09205606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980348 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980299 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980324 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09720453 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980322 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980293 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980315 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09227991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980368 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09407714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980317 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09246482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08982974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09364777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980392 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09187848 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09205602 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980308 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980339 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09720450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980379 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980404 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09682948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09730789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980328 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980292 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09615024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980330 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27980344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09187845 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09152326 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09040082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09035400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08982977 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10389336 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389332 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389337 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389333 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389338 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389334 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10389335 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452880_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10389331 | - | - |
| Mouse Genome 430 2.0 Array | 1452880_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452880_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 448850_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 448850_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100031352.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031352.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031352.mm.1 | TC1100003421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031354.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031354.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031354.mm.1 | TC1100003421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031355.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031355.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031355.mm.1 | TC1100003421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031356.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031356.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031356.mm.1 | TC1100003421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031358.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031358.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031358.mm.1 | TC1100003421.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100031353.mm.1 | NM_001005223.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031353.mm.1 | NM_001005223.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100031353.mm.1 | TC1100003421.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452880_at | - | - |
| Mouse Expression Set 430 | 1452880_PM_at | - | - |
| Murine Genome U74v2 Set | 160060_at | - | - |
| Murine Genome U74v2 Set | 160060_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452880_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452880_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452880_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 911061 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2457827 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2914957 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2914957 | - | - |

Znhit3
Genomic Map