Assay Details
| Gene Symbol: Aatf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019816.1 | NP_062790.1 | 9 - 10 | 1414 | 82 | |
| GenBank mRNA | AF222801.1 | - | 9 - 10 | 1414 | 82 | |
| AK016820.1 | - | 5 - 6 | 651 | 82 | ||
| BC025080.1 | - | 9 - 10 | 1401 | 2648117 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aatf
Genomic Map