Assay Details
| Gene Symbol: Lmbrd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177178.3 | NP_796152.2 | 6 - 7 | 969 | 77 | |
| GenBank mRNA | AK036527.1 | - | 6 - 7 | 990 | 77 | |
| AK079450.1 | - | 6 - 7 | 969 | 77 | ||
| AK153728.1 | - | 6 - 7 | 920 | 77 | ||
| BC095971.1 | - | 6 - 7 | 886 | 30108856 | 77 | |
| BC120727.1 | - | 6 - 7 | 941 | 40129650 | 77 | |
| BC125421.1 | - | 6 - 7 | 941 | 40129936 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lmbrd2
Genomic Map