Assay Details
| Gene Symbol: Nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040395.4 | NP_001035485.2 | 1 - 2 | 508 | 70 | |
| NM_001085410.2 | NP_001078879.1 | 1 - 2 | 508 | 70 | ||
| NM_001286253.1 | NP_001273182.1 | 1 - 2 | 508 | 70 | ||
| NM_001286255.1 | NP_001273184.1 | 1 - 2 | 508 | 70 | ||
| GenBank mRNA | AK016988.1 | - | 1 - 2 | 74 | 70 | |
| AK046666.1 | - | 1 - 2 | 509 | 70 | ||
| AK078318.1 | - | 1 - 2 | 111 | 70 | ||
| AK149267.1 | - | 1 - 2 | 88 | 70 | ||
| AK162601.1 | - | 1 - 2 | 82 | 70 | ||
| BC115776.1 | - | 1 - 2 | 87 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK149267.1 transcripts.
Set Membership: |
|

Nadk2
Genomic Map