Assay Details
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011845.2 | NP_035975.1 | 4 - 5 | 1035 | 77 | |
| GenBank mRNA | AF196480.1 | - | 4 - 5 | 1035 | 77 | |
| Y18881.1 | - | 5 - 6 | 1390 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mid2
Genomic Map