Assay Details
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011845.2 | NP_035975.1 | 5 - 6 | 1168 | 145 | |
| GenBank mRNA | AF196480.1 | - | 5 - 6 | 1168 | 145 | |
| AK039479.1 | - | 1 - 2 | 776 | 145 | ||
| Y18881.1 | - | 6 - 7 | 1523 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mid2
Genomic Map