Assay Details
| Gene Symbol: Mid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011845.2 | NP_035975.1 | 7 - 8 | 1470 | 107 | |
| GenBank mRNA | AF196480.1 | - | 7 - 8 | 1470 | 107 | |
| Y18881.1 | - | 8 - 9 | 1825 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mid2
Genomic Map