Assay Details
| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178309.2 | NP_840094.1 | 12 - 13 | 1918 | 142 | |
| GenBank mRNA | AK033205.1 | - | 12 - 13 | 1889 | 142 | |
| AK145709.1 | - | 12 - 13 | 1919 | 142 | ||
| AK169440.1 | - | 12 - 13 | 1865 | 142 | ||
| AK169823.1 | - | 11 - 12 | 1655 | 142 | ||
| BC094252.1 | - | 12 - 13 | 1867 | 6401109 | 142 | |
More Information
Updated Mapping Information
Set Membership: |
|

Brip1
Genomic Map