Assay Details
Gene Symbol: Brip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178309.2 | NP_840094.1 | 13 - 14 | 2056 | 60 | |
GenBank mRNA | AK033205.1 | - | 13 - 14 | 2027 | 60 | |
AK145709.1 | - | 13 - 14 | 2057 | 60 | ||
AK169440.1 | - | 13 - 14 | 2003 | 60 | ||
AK169823.1 | - | 12 - 13 | 1793 | 60 | ||
BC094252.1 | - | 13 - 14 | 2005 | 6401109 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
