Assay Details
| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178309.2 | NP_840094.1 | 1 - 2 | 91 | 80 | |
| GenBank mRNA | AK033205.1 | - | 1 - 2 | 63 | 80 | |
| AK051878.1 | - | 1 - 2 | 92 | 80 | ||
| AK145709.1 | - | 1 - 2 | 92 | 80 | ||
| AK169823.1 | - | 1 - 2 | 50 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK051878.1, AK145709.1, AK169823.1, NM_178309.2 transcripts.
Set Membership: |
|

Brip1
Genomic Map