Assay Details
| Gene Symbol: Brip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178309.2 | NP_840094.1 | 6 - 7 | 752 | 86 | |
| GenBank mRNA | AK033205.1 | - | 6 - 7 | 724 | 86 | |
| AK051878.1 | - | 6 - 7 | 753 | 86 | ||
| AK145709.1 | - | 6 - 7 | 753 | 86 | ||
| AK169440.1 | - | 6 - 7 | 699 | 86 | ||
| AK169823.1 | - | 6 - 7 | 711 | 86 | ||
| BC094252.1 | - | 6 - 7 | 701 | 6401109 | 86 | |
More Information
Set Membership: |
|

Brip1
Genomic Map