Assay Details
| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030688.1 | NP_109613.1 | 3 - 4 | 471 | 73 | |
| GenBank mRNA | AF284437.1 | - | 3 - 4 | 471 | 73 | |
| AJ277831.1 | - | 1229 | 73 | |||
| AK039155.1 | - | 996 | 73 | |||
| AK045976.1 | - | 1260 | 73 | |||
| BC119274.1 | - | 3 - 4 | 414 | 8734023 | 73 | |
| BC120555.1 | - | 3 - 4 | 414 | 40129478 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rapl2
Genomic Map