Assay Details
| Gene Symbol: Il1rapl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030688.1 | NP_109613.1 | 7 - 8 | 1021 | 101 | |
| GenBank mRNA | AF284437.1 | - | 7 - 8 | 1021 | 101 | |
| AJ277831.1 | - | 1779 | 101 | |||
| AK045976.1 | - | 1810 | 101 | |||
| BC119274.1 | - | 7 - 8 | 964 | 8734023 | 101 | |
| BC120555.1 | - | 7 - 8 | 964 | 40129478 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il1rapl2
Genomic Map