Assay Details
| Gene Symbol: B4galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008081.3 | NP_032107.1 | 1 - 2 | 58 | 56 | |
| GenBank mRNA | BC139165.1 | - | 1 - 2 | 48 | 8862187 | 56 |
| BC139166.1 | - | 1 - 2 | 48 | 8862188 | 56 | |
| L30104.1 | - | 1 - 2 | 58 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC139165.1, BC139166.1, L30104.1, NM_008081.3 transcripts.
Set Membership: |
|

B4galnt2
Genomic Map