Assay Details
| Gene Symbol: B4galnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008081.3 | NP_032107.1 | 5 - 6 | 555 | 58 | |
| GenBank mRNA | BC071231.1 | - | 5 - 6 | 523 | 58 | |
| BC139165.1 | - | 5 - 6 | 545 | 8862187 | 58 | |
| BC139166.1 | - | 5 - 6 | 545 | 8862188 | 58 | |
| L30104.1 | - | 5 - 6 | 555 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

B4galnt2
Genomic Map