Assay Details
| Gene Symbol: Egflam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289496.1 | NP_001276425.1 | 12 - 13 | 1995 | 73 | |
| NM_001289498.1 | NP_001276427.1 | 12 - 13 | 1990 | 73 | ||
| NM_178748.6 | NP_848863.1 | 12 - 13 | 1995 | 73 | ||
| GenBank mRNA | AK031136.1 | - | 12 - 13 | 1904 | 73 | |
| AK033332.1 | - | 12 - 13 | 1912 | 73 | ||
| AK037223.1 | - | 12 - 13 | 1897 | 73 | ||
| AK041546.1 | - | 12 - 13 | 1876 | 73 | ||
| AK087659.1 | - | 12 - 13 | 1875 | 73 | ||
| BC095994.1 | - | 12 - 13 | 1725 | 6467602 | 73 | |
| BC150710.1 | - | 12 - 13 | 1715 | 9087621 | 73 | |
| DQ223720.1 | - | 12 - 13 | 1699 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Egflam
Genomic Map