Assay Details
| Gene Symbol: Egflam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289496.1 | NP_001276425.1 | 2 - 3 | 508 | 67 | |
| NM_001289498.1 | NP_001276427.1 | 2 - 3 | 508 | 67 | ||
| NM_178748.6 | NP_848863.1 | 2 - 3 | 508 | 67 | ||
| GenBank mRNA | AK031136.1 | - | 2 - 3 | 417 | 67 | |
| AK033332.1 | - | 2 - 3 | 425 | 67 | ||
| AK037223.1 | - | 2 - 3 | 410 | 67 | ||
| AK041546.1 | - | 2 - 3 | 389 | 67 | ||
| AK087659.1 | - | 2 - 3 | 388 | 67 | ||
| AK158909.1 | - | 2 - 3 | 509 | 67 | ||
| BC095994.1 | - | 2 - 3 | 238 | 6467602 | 67 | |
| BC150710.1 | - | 2 - 3 | 228 | 9087621 | 67 | |
| DQ223720.1 | - | 2 - 3 | 212 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001289498.1 transcripts.
Set Membership: |
|

Egflam
Genomic Map