Assay Details
| Gene Symbol: Pigs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201406.1 | NP_958808.1 | 4 - 5 | 429 | 89 | |
| GenBank mRNA | AK033331.1 | - | 4 - 5 | 403 | 89 | |
| AK039604.1 | - | 4 - 5 | 400 | 89 | ||
| AK043232.1 | - | 4 - 5 | 403 | 89 | ||
| AK045759.1 | - | 4 - 5 | 400 | 89 | ||
| AK053771.1 | - | 4 - 5 | 403 | 89 | ||
| AK142871.1 | - | 4 - 5 | 400 | 89 | ||
| AK151858.1 | - | 4 - 5 | 391 | 89 | ||
| AK152229.1 | - | 4 - 5 | 391 | 89 | ||
| AK153300.1 | - | 4 - 5 | 395 | 89 | ||
| AK166408.1 | - | 4 - 5 | 406 | 89 | ||
| BC058979.1 | - | 4 - 5 | 429 | 5716747 | 89 | |
More Information
Set Membership: |
|

Pigs
Genomic Map