Assay Details
| Gene Symbol: Pigs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201406.1 | NP_958808.1 | 7 - 8 | 868 | 59 | |
| GenBank mRNA | AK033331.1 | - | 7 - 8 | 842 | 59 | |
| AK039604.1 | - | 7 - 8 | 839 | 59 | ||
| AK043232.1 | - | 7 - 8 | 842 | 59 | ||
| AK044292.1 | - | 1 - 2 | 464 | 59 | ||
| AK053771.1 | - | 7 - 8 | 842 | 59 | ||
| AK142871.1 | - | 7 - 8 | 839 | 59 | ||
| AK151858.1 | - | 7 - 8 | 830 | 59 | ||
| AK152229.1 | - | 7 - 8 | 830 | 59 | ||
| AK153300.1 | - | 7 - 8 | 834 | 59 | ||
| AK169633.1 | - | 6 - 7 | 641 | 59 | ||
| AK182014.1 | - | 2 - 3 | 158 | 59 | ||
| AK183805.1 | - | 1 - 2 | 158 | 59 | ||
| AK187604.1 | - | 1 - 2 | 158 | 59 | ||
| BC058979.1 | - | 7 - 8 | 868 | 5716747 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pigs
Genomic Map