Assay Details
| Gene Symbol: Morc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193309.1 | NP_001180238.1 | 13 - 14 | 1783 | 71 | |
| NM_029413.4 | NP_083689.2 | 13 - 14 | 1783 | 71 | ||
| GenBank mRNA | AK017472.1 | - | 13 - 14 | 1752 | 71 | |
| AK032807.1 | - | 13 - 14 | 1784 | 71 | ||
| BC036164.1 | - | 7 - 8 | 670 | 71 | ||
| BC039283.1 | - | 6 - 7 | 495 | 71 | ||
| BC098483.1 | - | 13 - 14 | 1738 | 30022418 | 71 | |
| BC132153.1 | - | 13 - 14 | 1637 | 40130430 | 71 | |
| BC132497.1 | - | 13 - 14 | 1637 | 40130774 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Morc4
Genomic Map