Assay Details
| Gene Symbol: Cdc20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281487.1 | NP_001268416.1 | 8 - 9 | 990 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc20b| Gene Symbol: Cdc20b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281487.1 | NP_001268416.1 | 8 - 9 | 990 | 58 | |
Set Membership: |
|