Assay Details
| Gene Symbol: 1700008I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027952.3 | NP_082228.2 | 5 - 6 | 642 | 108 | |
| GenBank mRNA | AK005772.1 | - | 5 - 6 | 643 | 108 | |
| BC141320.1 | - | 5 - 6 | 623 | 9055932 | 108 | |
| BC141322.1 | - | 5 - 6 | 623 | 9055934 | 108 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700008I05Rik
Genomic Map