Assay Details
| Gene Symbol: 1700008I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027952.3 | NP_082228.2 | 8 - 9 | 1186 | 121 | |
| GenBank mRNA | AK005772.1 | - | 8 - 9 | 1187 | 121 | |
| BC141320.1 | - | 8 - 9 | 1167 | 9055932 | 121 | |
| BC141322.1 | - | 8 - 9 | 1167 | 9055934 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 437512 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810533 | AK159290.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810533 | BC005474.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 810533 | NM_013901.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 370754 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787628 | NM_008936.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 787628 | U77946.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166018 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 911297 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 155091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701939 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160156 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275993 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | AF384162.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | AK002693.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | BC031920.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | BC106135.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | NM_026713.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 749505 | XM_001477283.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 824757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136496 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628274 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545561 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850254 | AK040679.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69842 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115025 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322495 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 348447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517745 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606354 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27073 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168708 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169403 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 127238 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P146627 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102041 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677900 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677906 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677910 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677911 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677896 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677897 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677901 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677904 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677913 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677898 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677908 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677909 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32677902 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09428557 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P146627 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09428492 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102041 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03758903 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03758936 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03758930 | - | - |
| Whole Mouse Genome G4122A | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | BC141322.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | NM_027952.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | AK005772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P200599 | BC141320.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P146627 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09428492 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102041 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975808 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975817 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975816 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975813 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975818 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14975806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677864 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677814 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677812 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677908 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677806 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677902 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677739 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677906 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677884 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677904 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677786 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677859 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677754 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677848 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677823 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677829 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32677810 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14975818 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677889 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677879 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14705358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677906 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677883 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677910 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677909 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677900 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677908 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677913 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15035304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677901 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677891 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677911 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677898 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14975806 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677882 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14845268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14975807 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14588665 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32677893 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10606825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10606830 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1447007_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453339_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10606820 | - | - |
| Mouse Genome 430 2.0 Array | 1447007_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453339_at | - | - |
| Mouse Genome 430 2.0 Array | 1453339_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1447007_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71841_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71841_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00018882.mm.1 | NM_027952.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018882.mm.1 | NM_027952.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00018882.mm.1 | TC0X00003014.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453339_PM_at | - | - |
| Mouse Expression Set 430 | 1453339_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447007_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453339_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1447007_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1453339_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 326745 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2676443 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2676443 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2676443 | - | - |

1700008I05Rik
Genomic Map