Assay Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172779.4 | NP_766367.1 | 12 - 13 | 1992 | 66 | |
| GenBank mRNA | AK083942.1 | - | 12 - 13 | 1954 | 66 | |
| AK144026.1 | - | 2 - 3 | 1837 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ddx26b
Genomic Map