Assay Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172779.4 | NP_766367.1 | 4 - 5 | 791 | 77 | |
| GenBank mRNA | AK031945.1 | - | 4 - 5 | 783 | 77 | |
| AK083942.1 | - | 4 - 5 | 753 | 77 | ||
| AK160627.1 | - | 4 - 5 | 751 | 77 | ||
| BC051161.1 | - | 4 - 5 | 773 | 6466142 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx26b
Genomic Map