Assay Details
| Gene Symbol: Ddx26b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172779.4 | NP_766367.1 | 5 - 6 | 977 | 150 | |
| GenBank mRNA | AK031945.1 | - | 5 - 6 | 969 | 150 | |
| AK083942.1 | - | 5 - 6 | 939 | 150 | ||
| AK160627.1 | - | 5 - 6 | 937 | 150 | ||
| BC051161.1 | - | 5 - 6 | 959 | 6466142 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx26b
Genomic Map