Assay Details
| Gene Symbol: Slc9a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172780.3 | NP_766368.2 | 4 - 5 | 705 | 71 | |
| GenBank mRNA | AK032326.1 | - | 4 - 5 | 686 | 71 | |
| AK122232.1 | - | 4 - 5 | 676 | 71 | ||
| BC130221.1 | - | 4 - 5 | 690 | 40141414 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a6
Genomic Map