Assay Details
| Gene Symbol: Slc38a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178746.4 | NP_848861.1 | 1 - 2 | 106 | 80 | |
| GenBank mRNA | AK034958.1 | - | 1 - 2 | 92 | 80 | |
| AK039821.1 | - | 1 - 2 | 91 | 80 | ||
| AK043402.1 | - | 1 - 2 | 94 | 80 | ||
| AK044735.1 | - | 1 - 2 | 96 | 80 | ||
| BC052361.1 | - | 1 - 2 | 106 | 6705789 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK043402.1, AK044735.1, BC052361.1, NM_178746.4 transcripts.
Set Membership: |
|

Slc38a9
Genomic Map