Assay Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079849.2 | NP_001073318.1 | 4 - 5 | 511 | 69 | |
| NM_145457.4 | NP_663432.1 | 4 - 5 | 729 | 69 | ||
| GenBank mRNA | AF128835.1 | - | 3 - 4 | 411 | 69 | |
| AK040012.1 | - | 4 - 5 | 512 | 69 | ||
| BB202059.2 | - | 1 - 2 | 100 | 69 | ||
| BC019726.1 | - | 4 - 5 | 700 | 4010170 | 69 | |
| BC051042.1 | - | 4 - 5 | 686 | 4008643 | 69 | |
| CB840007.1 | - | 3 - 4 | 484 | 69 | ||
| CD363907.1 | - | 4 - 5 | 487 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paip1
Genomic Map