Assay Details
| Gene Symbol: Paip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079849.2 | NP_001073318.1 | 5 - 6 | 623 | 127 | |
| NM_145457.4 | NP_663432.1 | 5 - 6 | 841 | 127 | ||
| GenBank mRNA | AF128835.1 | - | 4 - 5 | 523 | 127 | |
| AK040012.1 | - | 5 - 6 | 624 | 127 | ||
| BB202059.2 | - | 2 - 3 | 212 | 127 | ||
| BC019726.1 | - | 5 - 6 | 812 | 4010170 | 127 | |
| BC051042.1 | - | 5 - 6 | 798 | 4008643 | 127 | |
| CD363907.1 | - | 5 - 6 | 599 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paip1
Genomic Map