Assay Details
| Gene Symbol: Apobec3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB461446.1 | - | 1 - 2 | 49 | 87 | |
| AB461447.1 | - | 1 - 2 | 49 | 87 | ||
| AB461448.1 | - | 1 - 2 | 49 | 87 | ||
| AK044394.1 | - | 1 - 2 | 118 | 87 | ||
| AK154035.1 | - | 163 | 87 | |||
| BC003314.1 | - | 1 - 2 | 140 | 3155422 | 87 | |
| EU707568.1 | - | 1 - 2 | 49 | 87 | ||
| GQ871501.1 | - | 1 - 2 | 46 | 87 | ||
| GQ871503.1 | - | 1 - 2 | 46 | 87 | ||
More Information
Set Membership: |
|

Apobec3
Genomic Map