Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500005135.mm.1 |
NM_001160415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005135.mm.1 |
NM_001160415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005135.mm.1 |
NM_030255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005135.mm.1 |
NM_030255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005135.mm.1 |
TC1500000688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005133.mm.1 |
NM_001160415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005133.mm.1 |
NM_001160415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005133.mm.1 |
NM_030255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005133.mm.1 |
NM_030255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005133.mm.1 |
TC1500000688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005130.mm.1 |
NM_001160415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005130.mm.1 |
NM_001160415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005130.mm.1 |
NM_030255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005130.mm.1 |
NM_030255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005130.mm.1 |
TC1500000688.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500005137.mm.1 |
NM_001160415.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005137.mm.1 |
NM_001160415.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005137.mm.1 |
NM_030255.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005137.mm.1 |
NM_030255.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500005137.mm.1 |
TC1500000688.mm.1 |
MTA TC |