Assay Details
| Gene Symbol: Rpl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013762.2 | NP_038790.2 | 1 - 2 | 87 | 58 | |
| GenBank mRNA | AK088993.1 | - | 1 - 2 | 34 | 58 | |
| AK144518.1 | - | 1 - 2 | 34 | 58 | ||
| AK144816.1 | - | 1 - 2 | 34 | 58 | ||
| AK146502.1 | - | 1 - 2 | 34 | 58 | ||
| AK149494.1 | - | 1 - 2 | 34 | 58 | ||
| AK151057.1 | - | 1 - 2 | 34 | 58 | ||
| AK151084.1 | - | 1 - 2 | 34 | 58 | ||
| AK151142.1 | - | 1 - 2 | 34 | 58 | ||
| AK151426.1 | - | 1 - 2 | 34 | 58 | ||
| AK151596.1 | - | 1 - 2 | 34 | 58 | ||
| AK151740.1 | - | 1 - 2 | 34 | 58 | ||
| AK166825.1 | - | 1 - 2 | 34 | 58 | ||
| AK167787.1 | - | 1 - 2 | 34 | 58 | ||
| AK167947.1 | - | 1 - 2 | 34 | 58 | ||
| AK168040.1 | - | 1 - 2 | 34 | 58 | ||
| AK168059.1 | - | 1 - 2 | 33 | 58 | ||
| AK168128.1 | - | 1 - 2 | 34 | 58 | ||
| AK169060.1 | - | 1 - 2 | 34 | 58 | ||
| AK170819.1 | - | 1 - 2 | 34 | 58 | ||
| AK172280.1 | - | 1 - 2 | 34 | 58 | ||
| BC158039.1 | - | 1 - 2 | 34 | 9007342 | 58 | |
| CJ042214.1 | - | 1 - 2 | 87 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK088993.1, AK144518.1, AK144816.1, AK146502.1, AK149494.1, AK151057.1, AK151084.1, AK151142.1, AK151426.1, AK151596.1, AK151740.1, AK166825.1, AK167787.1, AK167947.1, AK168040.1, AK168059.1, AK168128.1, AK169060.1, AK170819.1, AK172280.1, BC158039.1, NM_013762.2 transcripts.
Set Membership: |
|

Rpl3
Genomic Map