Assay Details
| Gene Symbol: Grap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289442.1 | NP_001276371.1 | 4 - 5 | 445 | 69 | |
| NM_010815.3 | NP_034945.1 | 3 - 4 | 540 | 69 | ||
| GenBank mRNA | AF053405.1 | - | 3 - 4 | 508 | 69 | |
| AF055465.1 | - | 3 - 4 | 540 | 69 | ||
| AF129477.1 | - | 3 - 4 | 503 | 69 | ||
| AF236118.1 | - | 3 - 4 | 510 | 69 | ||
| AJ011735.1 | - | 3 - 4 | 506 | 69 | ||
| AK156939.1 | - | 3 - 4 | 519 | 69 | ||
| BC034859.1 | - | 4 - 5 | 682 | 69 | ||
| BC052496.1 | - | 4 - 5 | 457 | 1446918 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grap2
Genomic Map