Assay Details
| Gene Symbol: Tab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025609.2 | NP_079885.2 | 2 - 3 | 164 | 82 | |
| GenBank mRNA | AB088136.1 | - | 2 - 3 | 164 | 82 | |
| BC027054.1 | - | 2 - 3 | 156 | 82 | ||
| BC041110.1 | - | 2 - 3 | 159 | 82 | ||
| BC054369.1 | - | 2 - 3 | 164 | 5356886 | 82 | |
| BC114637.1 | - | 2 - 3 | 185 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tab1
Genomic Map