Assay Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113386.1 | NP_001106857.1 | 13 - 14 | 2069 | 123 | |
| NM_013584.2 | NP_038612.1 | 12 - 13 | 1718 | 123 | ||
| GenBank mRNA | AK155524.1 | - | 12 - 13 | 1945 | 123 | |
| BC031929.1 | - | 12 - 13 | 1711 | 4159053 | 123 | |
| D17444.1 | - | 12 - 13 | 1725 | 123 | ||
| D26177.1 | - | 12 - 13 | 1725 | 123 | ||
| S73495.1 | - | 12 - 13 | 1725 | 123 | ||
| S73496.1 | - | 12 - 13 | 1725 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lifr
Genomic Map