Assay Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113386.1 | NP_001106857.1 | 6 - 7 | 958 | 89 | |
| NM_013584.2 | NP_038612.1 | 5 - 6 | 607 | 89 | ||
| GenBank mRNA | AK145864.1 | - | 6 - 7 | 959 | 89 | |
| AK155524.1 | - | 5 - 6 | 834 | 89 | ||
| BC031929.1 | - | 5 - 6 | 600 | 4159053 | 89 | |
| D17444.1 | - | 5 - 6 | 614 | 89 | ||
| D26177.1 | - | 5 - 6 | 614 | 89 | ||
| S73495.1 | - | 5 - 6 | 614 | 89 | ||
| S73496.1 | - | 5 - 6 | 614 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lifr
Genomic Map