Assay Details
| Gene Symbol: Lifr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113386.1 | NP_001106857.1 | 7 - 8 | 1132 | 76 | |
| NM_013584.2 | NP_038612.1 | 6 - 7 | 781 | 76 | ||
| GenBank mRNA | AK145864.1 | - | 7 - 8 | 1133 | 76 | |
| AK155524.1 | - | 6 - 7 | 1008 | 76 | ||
| BC031929.1 | - | 6 - 7 | 774 | 4159053 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lifr
Genomic Map