Assay Details
| Gene Symbol: Rad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289448.1 | NP_001276377.1 | 1 - 2 | 245 | 73 | |
| NM_011232.3 | NP_035362.2 | 1 - 2 | 245 | 73 | ||
| GenBank mRNA | AF038841.1 | - | 1 - 2 | 156 | 73 | |
| AF073523.1 | - | 1 - 2 | 172 | 73 | ||
| AF084514.1 | - | 1 - 2 | 104 | 73 | ||
| AK146533.1 | - | 1 - 2 | 215 | 73 | ||
| AK148098.1 | - | 1 - 2 | 204 | 73 | ||
| AK150466.1 | - | 1 - 2 | 154 | 73 | ||
| AK168588.1 | - | 1 - 2 | 195 | 73 | ||
| BX517768.1 | - | 1 - 2 | 230 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF038841.1, AF073523.1, AF084514.1, AK146533.1, AK148098.1, AK150466.1, AK168588.1, NM_001289448.1, NM_011232.3 transcripts.
Set Membership: |
|

Rad1
Genomic Map