Assay Details
| Gene Symbol: Rad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289447.1 | NP_001276376.1 | 3 - 4 | 407 | 145 | |
| NM_001289448.1 | NP_001276377.1 | 3 - 4 | 619 | 145 | ||
| NM_011232.3 | NP_035362.2 | 3 - 4 | 619 | 145 | ||
| GenBank mRNA | AF038841.1 | - | 3 - 4 | 530 | 145 | |
| AF058394.1 | - | 2 - 3 | 306 | 145 | ||
| AF073523.1 | - | 3 - 4 | 546 | 145 | ||
| AF074718.1 | - | 2 - 3 | 316 | 145 | ||
| AF076842.1 | - | 2 - 3 | 317 | 145 | ||
| AF084514.1 | - | 3 - 4 | 478 | 145 | ||
| AJ004976.1 | - | 2 - 3 | 328 | 145 | ||
| AK146533.1 | - | 3 - 4 | 589 | 145 | ||
| AK150466.1 | - | 3 - 4 | 528 | 145 | ||
| AK164168.1 | - | 2 - 3 | 777 | 145 | ||
| AK168588.1 | - | 3 - 4 | 569 | 145 | ||
| BC048693.1 | - | 2 - 3 | 383 | 6508142 | 145 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad1
Genomic Map