Assay Details
| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008942.2 | NP_032968.2 | 20 - 21 | 2524 | 91 | |
| GenBank mRNA | AK133898.1 | - | 20 - 21 | 2332 | 91 | |
| BC009653.1 | - | 20 - 21 | 2463 | 3487224 | 91 | |
| BC086798.1 | - | 20 - 21 | 2516 | 6812900 | 91 | |
| BC098212.1 | - | 20 - 21 | 2524 | 6825181 | 91 | |
| U35646.1 | - | 20 - 21 | 2511 | 91 | ||
More Information
Set Membership: |
|

Npepps
Genomic Map