Assay Details
| Gene Symbol: Npepps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008942.2 | NP_032968.2 | 7 - 8 | 1069 | 129 | |
| GenBank mRNA | AK133898.1 | - | 7 - 8 | 877 | 129 | |
| BC009653.1 | - | 7 - 8 | 1008 | 3487224 | 129 | |
| BC086798.1 | - | 7 - 8 | 1061 | 6812900 | 129 | |
| BC098212.1 | - | 7 - 8 | 1069 | 6825181 | 129 | |
| U35646.1 | - | 7 - 8 | 1056 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npepps
Genomic Map