Assay Details
| Gene Symbol: Larp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024526.2 | NP_001019697.2 | 11 - 12 | 1505 | 80 | |
| NM_001080948.2 | NP_001074417.1 | 11 - 12 | 1518 | 80 | ||
| NM_001284521.1 | NP_001271450.1 | 11 - 12 | 1492 | 80 | ||
| NM_001284522.1 | NP_001271451.1 | 11 - 12 | 1515 | 80 | ||
| NM_001284523.1 | NP_001271452.1 | 11 - 12 | 1508 | 80 | ||
| GenBank mRNA | AK049756.1 | - | 11 - 12 | 1432 | 80 | |
| AK139636.1 | - | 11 - 12 | 1513 | 80 | ||
| AK139909.1 | - | 11 - 12 | 1356 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Larp4
Genomic Map